Learn More
Invitrogen™ Progesterone Receptor Monoclonal Antibody (Alpha PR6)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA1411
Descrizione
MA1-411 reacts with high affinity to the B form of the progesterone receptor from human, mouse, rat, rabbit, bovine, ovine, chicken, and guinea pig tissues. This antibody does not cross-react with estrogen receptor or glucocorticoid receptor. MA1-411 has been successfully used in Western blot, immunocytochemical, immunohistochemical and immunoprecipitation procedures. By Western blot, this antibody detects a 110 kDa protein representing the B form of the PR in chickens.' The MA1-411 immunogen is progesterone receptor purified from chick oviduct cytosol. Reconstitute with PBS.
The progesterone receptor (PR) is a member of the steroid family of nuclear receptors. The PR mediates the physiological effects of progesterone, which plays a central role in reproductive events associated with the establishment and maintenance of pregnancy. PR is found as a 94 kDa protein (Form A) or a 120 kDa protein (Form B) due to the use of alternative translation initiation sites. PR-B is the transcriptionally active form and is responsible for activating genes for the maintenance of the endometrium, maintenance of pregnancy, and inhibition of ovulation. PR-A is identical to PR-B except for a 165 amino acid deletion at the N-terminus. This deletion exposes a 140 amino acid inhibitory domain (ID) that acts as a repressor of steroid hormone transcriptional activity. In its inactive state, PgR forms a multiprotein complex which includes heat shock proteins and immunophins. Upon binding of progesterone hormone to its receptor, there is a conformational change that allows dimerization and binding of the receptor to progesterone response elements (PRE) sequences, resulting in activated transcription. A Null mutation in the PGR gene leads to pleiotrophic reproductive abnormalities.
Specifica
| Progesterone Receptor | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P06186, P06401, P07812, Q00175, Q28590, Q63449 | |
| PGR | |
| Progesterone receptor purified from chick oviduct cytosol. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Rabbit, Bovine, Sheep, Chicken, Guinea Pig | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Alpha PR6 | |
| Unconjugated | |
| PGR | |
| 9930019P03Rik; BB114106; ENSMUSG00000074510; LOW QUALITY PROTEIN: progesterone receptor; NR3C3; Nuclear receptor subfamily 3 group C member 3; p4 receptor; PGR; pot. alt. progesterone receptor; putative; PR; PR-A; PR-B; Progesterone receptor; progesterone receptor A; progesterone-receptor; putative form B; steroid hormone receptor; testicular progesterone receptor; unnamed protein product | |
| Mouse | |
| Protein A | |
| RUO | |
| 100009094, 100724945, 100848466, 18667, 25154, 396198, 443444, 5241 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.