missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Prolactin Monoclonal Antibody (INN-hPRL-1)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA125051
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This product does require antigen retrieval using heat treatment (1mm EDTA, pH 8.0, is recommended) prior to staining of paraffin sections. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Prolactin (PRL) is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. PRL exerts its influence on different cell types through a signal transduction pathway which begins with the binding of the hormone to a transmembrane PRL receptor.
Specifica
| Prolactin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| P01236 | |
| PRL | |
| proprietary immunogen. | |
| 250 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Radioimmune Assays (RIA) | |
| INN-hPRL-1 | |
| Unconjugated | |
| PRL | |
| AV290867; decidual prolactin; GHA1; growth hormone A1; Human prolactin; lactogenic prehormone (AA -30 to 199); preprolactin; pre-prolactin; Prl; Prl1a1; PRLB; PRLSD1; Prol; Prolactin; prolactin family 1, subfamily a, member 1; prolactin precursor; prolactin protein; RATPRLSD1; RNPROL | |
| Mouse | |
| Ion-exchange Chromatography | |
| RUO | |
| 5617 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto