Learn More
DiscoverX Prolactin Releasing Peptide (1-31)
Click to view available options
Quantità:
0.5 mg
Descrizione
Prolactin Releasing Peptide is a hypothalamic peptide hormone that stimulates prolactin release, and regulates prolactin expression, by binding to prolactin releasing peptide receptor PRLHR (GPR10). It has stimulatory effects on both luteinizing hormone and follicle stimulating hormone. Provided as 500μg of dry powder alongside reconstitution buffer.
Specifica
Specifica
| Product Type | Control Ligands |
| Quantità | 0.5 mg |