Learn More
Thermo Scientific™ ProPac™ PA1 HPLC Columns
Descrizione
Efficiently analyze and purify hydrophilic anionic proteins and peptides with Thermo Scientific™ ProPac™ PA1 LC Columns. These strong anion-exchange columns provide high-resolution separations of proteins with pI values from 3 to 11.
The columns can also resolve some proteins and peptides that are highly cationic and have very high isoelectric points.
- Good for hydrophilic proteins and peptides
- Ideal for high-resolution separations of proteins with pI values from 3 to 11
- Pellicular packing ensures high-efficiency and fast mass transport
Specifica
Specifica
| Formato colonna | Guard Column |
| Diametro (sistema metrico) | 4 mm |
| Portata | 0.9 to 1.5 mL/min |
| Da utilizzare con | BioLC Chromatography System |
| Lunghezza (sistema metrico) | 50 mm |
| Pressione max | 5000 psi (345 bar) |
| Dimensione particelle | 10 μm |
| Dimensione del poro | Nonporous |
| Quantità | Each |
| Fase stazionaria | Alkyl Quaternary Amine |
| Vedi altri risultati |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.