Learn More
Cytiva Whatman™ Protein Prep for ӒKTA™ Chromatography Systems
Codice prodotto. 15871838 Sfoglia Tutto Cytiva Prodotti
Click to view available options
Dimensione del poro:
0.2 μm
0.45 μm
Diametro (sistema metrico):
13 mm
30 mm
Dimensione della confezione:
pezzi 150
Codice prodotto. 15871838
Marca: Cytiva 10463113
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
The Protein Prep syringe filter containing the regenerated cellulose membrane is the optimal choice for filtration prior to chromatography with ӒKTA™ systems.
  • Suitable for numerous applications in pharmaceutical, environmental, biotechnology, food/beverage and agricultural testing laboratories
  • Heat sealed without the use of glues or sealants
  • Designed for optimal preparation of protein samples prior to chromatography
  • High yield protein purification and accurate protein analysis both depend on a quality sample being introduced to the chromatography column. The Protein Prep syringe filter has been designed specifically to optimize these outcomes
  • Filtration of samples prior to runs demonstrated to reduce column degrading particulate matter
  • Hydrophilic membrane made of regenerated cellulose exhibits low protein binding for sample preservation
  • Lot-to-lot certified to have low levels of extractables that could otherwise interfere with work
  • TRUSTED_SUSTAINABILITY
    Specifica
    Dimensione del poro 0.45 μm
    Imballaggio 150 Pieces
    Diametro (sistema metrico) 13 mm