Learn More
Cytiva Whatman™ Protein Prep for ӒKTA™ Chromatography Systems
Shop All Cytiva Products
Click to view available options
Dimensione del poro:
0.2 μm
0.45 μm
Diametro (sistema metrico):
13 mm
30 mm
Unit Size:
pezzi 150
Description
Specifications
Specifications
| Dimensione del poro | 0.2 μm |
| Imballaggio | 150 Pieces |
| Diametro (sistema metrico) | 30 mm |