Learn More
PROX1 Mouse anti-Human, Unconjugated, Clone: 1D4B1, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 67438-1-IG-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The PROX1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PROX1. This antibody recognizes human, rat, mouse antigen. The PROX1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifica
| PROX1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, ELISA | |
| 1D4B1 | |
| Unconjugated | |
| 20 μL | |
| 5629 | |
| IgG2b |