Learn More
Invitrogen™ Pseudomonas aeruginosa oprF Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5117553
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This antibody has been tested in direct-ELISA.
Has porin activity, forming small water-filled channels. Also has a structural role in determining cell shape and ability to grow in low-osmolarity medium.
Specifica
| Pseudomonas aeruginosa oprF | |
| Polyclonal | |
| Unconjugated | |
| oprF | |
| oprF | |
| Recombinant Pseudomonas aeruginosa Outer membrane porin F protein (25-350aa). | |
| 100 μL | |
| Primary | |
| Bacteria | |
| Antibody | |
| IgG |
| Western Blot | |
| 3.018 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P13794 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 878442 | |
| -20°C or -80°C if preferred | |
| Liquid |