Learn More
Aalborg™ PTFE Aluminum Shell Needle Valve
Codice prodotto. p-8490016
Click to view available options
Include:
1/8 in. NPTF Fitting
Glass Nipple Fitting
Dimensione della confezione:
Pezzo
Codice prodotto. 11820140
Marca: Aalborg™ VCHTT0G
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Aalborg™ PTFE Aluminum Shell Needle Valve is designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
  • Fluids in contact with PTFE only
  • Structurally rigid metal shell.
  • One PTFE o-ring
  • Simplicity - only seven components.
  • Up to 55 SLPM (air) or 2.8 LPM (water)
Specifica
Tipo Needle Valve
Include Glass Nipple Fitting
Velocità di flusso dell'aria 55 L/min.
Portata flusso d'acqua 28 mL/min.
Temperatura operativa max (sistema metrico) 65°C