Learn More
PTFE Beaker Cover-Watch Glass;Dia (mm), 80;Fits Beakers, 250ml
Codice prodotto. 15466175
Click to view available options
Descrizione:
Watch glass; Fits 1000mL Beakers
Watch glass; Fits 100mL Beakers
Watch glass; Fits 2000mL Beakers
Watch glass; Fits 250mL Beakers
Watch glass; Fits 25mL Beakers
Watch glass; Fits 5-10mL Beakers
Watch glass; Fits 500mL Beakers
Watch glass; Fits 50mL Beakers
Dimensione della confezione:
Pezzo
Codice prodotto. 15466175
Marca: Thermo Scientific Alfa Aesar 038096.KT
Solo null disponibili
Aggiungi al carrello
For use as a beaker cover or a watch glass
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Available in a range of diameters
TRUSTED_SUSTAINABILITY
Specifica
Descrizione Watch glass; Fits 250mL Beakers
Materiale PTFE
Tipo Becher