Learn More
DWK Life Sciences PTFE Hose Connection, with silicone seal (VMQ), complete with screw cap PBT, bent
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Nessun giunto rastremato standard:
GL 14
GL 18
Unit Size:
pezzi 10
Description
Hoses can be securely connected to laboratory apparatus using plastic nozzles screwed onto a glass thread using a screw cap with bore. Plastic nozzles made of PTFE for use with higher thermal and chemical resistance requirements.
- Hose olives enable the connection of hoses to laboratory apparatuses
Specifications
Specifications
| Quantità | 10/Pk. |
| Tipo | Bent |
| Joint Size | 9 mm |
| Nessun giunto rastremato standard | GL 14 |