Learn More
HUBER Drain Hose, Pressureless, T-180-NW8, PTFE
Click to view available options
:
Width: 0.8cm
Specifica
Specifica
| Materiale | PTFE |
| Min. Operating Temperature (Metric) | -60°C |
| Quantità confezione | 1 Ea. |
| Max. Operating Temperature (Metric) | 180°C |
| Product Type | Hose T-180-NW8 |