Learn More
Cowie Technology™ PTFE Joint Adapter
Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
N. giunto rastremato standard:
10/19
19/26
29/42
35/45
Cone Size:
14/23
24/29
29/32
45/50
Dimensione della confezione:
Pezzo
Descrizione
- Made from PTFE with a substantial shoulder for ease of handling
- Overall robust construction for repeated use
- Rigid sleeves and adapters are inert, usable to 280°C (approx.)
- Withstands high vacuum
- Transverse trapezoidal sealing rings on the contacting surfaces create high vacuum
Specifica
Specifica
| Cone Size | 45/50 |
| Product Type | PTFE Joint Adapters |
| Quantità | 1 Each |
| N. giunto rastremato standard | 35/45 |