Learn More
DWK Life Sciences PTFE Key with Thread, with three part retaining device
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Dimensione chiusura:
12.5 NS
14.5 NS
18.8 NS
21.5 NS
24 NS
29.2 NS
Foro spina:
2 mm
2.5 mm
4 mm
6 mm
8 mm
10 mm
Dimensione della confezione:
Pezzo
pezzi 10
pezzi 25
pezzi 50
Descrizione
Disposable PTFE valve plugs in combination with the polished DURAN™ disposable valve represent a grease-free alternative. The 3-part tap safety device is already included in the scope of delivery.
- Made of PTFE with thread and 3-part valve locking device
- For use with polished DURAN™ one-way taps
- Grease-free working
Specifica
Specifica
| Foro spina | 6 mm |
| Materiale | PTFE |
| Autoclavabile | Not Autoclavable |
| Dimensione chiusura | 18.8 NS |
| N. per confezione | 25/Pk. |
| Da utilizzare con (applicazione) | For use with polished DURAN™ one-way taps |