Learn More
Bohlender™ PTFE and PPS HT Laboratory Screw Joints, GL 18
Click to view available options
Dimensioni dei tubi:
2.0 mm O.D.
3.2 mm O.D.
6.35 mm O.D.
9.52 mm O.D.
Dimensione della confezione:
Pezzo
Descrizione
- Providing temperature resistance from −50° to +250°C
- Universal chemical resistance
- FDA conformity
- Max. pressure 10 bar
- Vacuum suitable
- Black screw cap made of PPS
- Inner parts consisting of a v-ring (PPS), a tapered ring and a sealing ring (both PTFE) as well as an o-ring (FKM) for tubing diameters under 3 mm
- Applications include fixing probes, thermometers, dip tubes or cables in reaction vessels
- Available for a wide range of GL thread sizes and tubing diameters
Specifica
Specifica
| Certificazione/Conformità | FDA |
| Materiale | PPS, PTFE |
| Da utilizzare con (applicazione) | Connecting equipment/fittings to hard-walled tubing |
| Min. Operating Temperature (Metric) | ° |
| Dimensioni dei tubi | 9.52 mm O.D. |
| Dimensioni filo | GL 18 |
| Pressione massima | 10 bar |
| Max. Operating Temperature (Metric) | 250°C |
| Product Type | Laboratory Screw Joint |