Learn More
Lenz Laborglasintrumente PTFE Stopcock Plugs
Codice prodotto. 10597190
Click to view available options
Foro spina:
1.5 mm
8 mm
Dimensione della confezione:
Pezzo
Codice prodotto. 10597190
Marca: Lenz Laborglasintrumente 02172408
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • PTFE stopcock plugs
  • With thread and screwthread retaining device
  • For stopcocks
  • Straight bore
  • Material: PTFE
Specifica
Foro spina 8 mm
Materiale PTFE