Learn More
Lenz Laborglasintrumente PTFE Stopcock Plugs
Codice prodotto. p-7222641
Click to view available options
Foro spina:
1.5 mm
8 mm
Dimensione della confezione:
Pezzo
Codice prodotto. 10077322
Marca: Lenz Laborglasintrumente 02171201
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • PTFE stopcock plugs
  • With thread and screwthread retaining device
  • For stopcocks
  • Straight bore
  • Material: PTFE
Specifica
Foro spina 1.5 mm
Materiale PTFE