Learn More
Adtech Polymer Engineering™ Tubo in PTFE
Click to view available options
Diametro (sistema metrico) interno:
0.5 mm
0.8 mm
4.8 mm
7.6 mm
Diametro (sistema metrico) esterno:
1.6 mm
6.4 mm
9.6 mm
Overall Length (Metric):
20 m
3.6 m
Dimensione della confezione:
Pezzo
Descrizione
- Non-stick characteristics
- Low friction surface
- Very high resistance to chemicals and solvents
- Very high electrical resistance
- Physiologically neutral
- Non-polluting and odorless
- Sterilization achieved through autoclaving, ethylene oxide or dry heat
Specifica
Specifica
| Autoclavabile | Autoclavable |
| Diametro (sistema metrico) interno | 0.8 mm |
| Materiale | PTFE |
| Diametro (sistema metrico) esterno | 1.6 mm |
| Overall Length (Metric) | 20 m |
| Min. Operating Temperature (Metric) | 260 °C |
| Quantità confezione | 1 Pack |
| Colore | Semitrasparente |
| Max. Operating Temperature (Metric) | 260 °C |