Learn More
Labconco™ Purifier™ Logic™+ Class II A2 Biosafety Cabinets, 4 ft. Width, International Models
Codice prodotto. p-7159740 Sfoglia Tutto Labconco Corporation Prodotti
Click to view available options
Tipo di spina:
Schuko (230 V)
Apertura del telaio scorrevole (sistema imperiale):
8 in.
Include:
Cabinet, Accessory Package, Base Stand
Dimensione della confezione:
Pezzo
Codice prodotto. 16645072
Marca: Labconco™ 302481131
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality
All models feature:
  • Electronically Commutated Motor (ECM)
  • Constant Airflow Profile (CAP) airflow monitoring system
  • Intrinsically-safe negative pressure design
  • Air-Wave™ Entry System
  • Contain-Air™ Negative Pressure Channel
  • Supply and exhaust 99.99% efficient HEPA filters. 99.999% efficient ULPA filters available
  • Interior-mounted, line-of-sight color LCD with MyLogic OS. Displays filter life, status messages for alarm conditions and alerts. 8 languages available
  • Bright, 90-150 footcandle, glare-free LED lighting located outside the contaminated work area
  • Fully-closing, clear 1/4 in. tempered safety glass sash
  • Curved stainless steel inlet grille with Reserve-Air Secondary Airflow Slots
  • Two electrical duplex receptacles. Flush-mounted-stainless steel with dampened hinges. Single outlets on 230 V models. One outlet on each side, with ground fault interruption (115 V)
  • Smart-Start™ System user-programmable start up and shut down
  • Night-Smart™ System reduces blower speed when sash is closed
  • Leak-tight stainless steel interior and powder-coated steel exterior
  • Removable towel catch located under work surface
  • Code-activated electronic security lock
  • 22.6 in. (57 cm) max. sash opening and 27 in. (69 cm) viewing height
  • 10° Angled sash with counterbalanced, easy lift mechanism
  • ADA-Compliant touchpad control on right side post
  • Noise level <63 dBA
  • Heat load 990 BTU/hour (4 ft. models)
  • Removable, seamless type 304 stainless steel, dished work surface with lift out knobs
  • Nominal inflow velocity of 105 fpm (0.5 m/sec)
  • Nominal downflow velocity of 55 fpm (0.3 m/sec)
  • Approximately 70% air recirculation
  • Overall depth x height: 31.2 x 61.7 in. (79.2 x 156.7 cm)
  • 10 ft (3 m) power cord with plug
  • Built-in timer and digital clock
  • Five year warranty
  • NSF International listed
  • Optional NSF-approved non-welded telescoping base stand included on some models
  • Optional Accessory Package (254 nm UV lamp, ADA-compliant service fixture(s) with ball-type valve(s), and NSF-Approved Vacu-Pass™ Cord & Cable Portal) included on some models
Specifications
Certificazione/Conformità NSF, CE Marked, ADA, ISO 5 Conditions per ISO 14644-1 and 2
Materiale camera Stainless Steel
Larghezza (sistema metrico) interno 123.2 cm
Product Type Type A2 Biosafety Cabinet
Modello Purifier Logic+
Livello di rumorosità <63 dBA
Prese 2
Tipo di spina Schuko (230 V)
Consumo energetico 270 W
Altezza (sistema imperiale) 89.2 to 95.2 in.
Apertura del telaio scorrevole (sistema metrico) 20.3 cm
Larghezza (sistema imperiale) parte esterna 54.3 in.
Volume (sistema metrico) di scarico 0.127 to 0.140 m3sec.
Larghezza (sistema metrico) parte esterna 137.922 cm
Include Cabinet, Accessory Package, Base Stand
Lighting LED, UV
Altezza (sistema metrico) 226.6 to 241.8 cm
Apertura del telaio scorrevole (sistema imperiale) 8 in.
Sistemi di manutenzione One
Warranty 5 Years
Larghezza (sistema imperiale) interno 48.5 in.
Larghezza (sistema metrico) 137.922 cm
Frequenza 50/60 Hz
Profondità (sistema imperiale) esterno 31.2 in.
Profondità (sistema metrico) esterno 79.248 cm
Requisiti elettrici 230 V, 50/60 Hz, 6 A
Tensione 230 V