Learn More
Labconco™ Purifier™ Logic™+ Class II B2 Biosafety Cabinets with Base Stand, International Models
Codice prodotto. 16629183 Sfoglia Tutto Labconco Corporation Prodotti
Click to view available options
Tipo di spina:
Schuko (230 V)
Larghezza (sistema imperiale) interno:
4 ft.
Include:
With Accessory Package
Dimensione della confezione:
Pezzo
Codice prodotto. 16629183
Marca: Labconco™ 303481131
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Total exhaust biohazard cabinets with advanced ergonomic design that excels in user comfort and functionality.
  • Electronically Commutated Motor (ECM)
  • Constant Airflow Profile (CAP) airflow monitoring system
  • Intrinsically-safe negative pressure design
  • Air-Wave Entry System
  • Contain-Air Negative Pressure Channel
  • Supply and exhaust 99.99% efficient HEPA filters. 99.999% efficient ULPA filters available
  • Interior-mounted, line-of-sight, color LCD with MyLogic OS displays filter life, status messages for alarm conditions and alerts. 8 languages available.
  • Bright, 90-150 footcandle, glare-free LED lighting located outside the contaminated work area
  • Fully-closing, clear tempered safety glass sash
  • Curved stainless steel inlet grille with Reserve-Air Secondary Airflow Slots
  • Two electrical duplex receptacles. Flush-mounted-stainless steel with dampened hinges. Single outlets on 230 V models. One outlet on each side, with ground fault interruption (115 V)
  • Smart-Start System user-programmable start up and shut down
  • Leak-tight stainless steel interior and powder-coated steel exterior
  • Removable towel catch located under work surface
  • Code-activated electronic security lock
  • 10° Angled sash with counterbalanced, easy lift mechanism
  • ADA-Compliant touchpad control on right side post
  • Removable, seamless type 304 stainless steel, dished work surface with lift out knobs
  • Heat load 990 BTU/hour (4 ft. models)
  • Nominal inflow velocity of 105 fpm (0.5 m/sec)
  • Nominal downflow velocity of 55 fpm (0.3 m/sec)
  • 10 ft. (3 m) power cord with plug
  • Five year warranty
  • NSF International listed
  • Optional NSF-approved non-welded telescoping base stand included on some models
  • Optional Accessory Package (254 nm UV lamp, ADA-compliant service fixture(s) with ball-type valve(s), and NSF-Approved Vacu-Pass Cord & Cable Portal) included on some models
  • Supporting base required if non-welded stand model is not selected
Specifica
Certificazione/Conformità ADA, CE, NSF 49
Materiale camera Stainless Steel
Larghezza (sistema metrico) interno 123.2 cm
Product Type Type B2 Biosafety Cabinet
Modello Purifier Logic+
Livello di rumorosità <63 dBA
Prese 2
Tipo di spina Schuko (230 V)
Consumo energetico 170 W
Altezza (sistema imperiale) 100.1 to 106.1 in.
Apertura del telaio scorrevole (sistema metrico) 20.3 cm
Larghezza (sistema imperiale) parte esterna 4.5 ft.
Volume (sistema metrico) di scarico 0.382 m3sec.
Larghezza (sistema metrico) parte esterna 137.92 cm
Include With Accessory Package
Lighting LED, UV
Altezza (sistema metrico) 254.2 to 269.2 cm
Apertura del telaio scorrevole (sistema imperiale) 8 in.
Sistemi di manutenzione One
Warranty 5 Years
Larghezza (sistema imperiale) interno 4 ft.
Profondità (sistema imperiale) esterno 31.2 in.
Profondità (sistema metrico) esterno 79.25 cm
Requisiti elettrici 230 V, 50/60 Hz, 6 A