Learn More
Ismatec™ PVC Solvent Ready Microbore Extension Tubing
Codice prodotto. p-8013010
Click to view available options
Dimensioni dei tubi:
26
42
48
Dimensione della confezione:
1 rotolo
15m
Codice prodotto. 12458620
Marca: Ismatec™ 9571226
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Specifically designed for use with hydrocarbons, petroleum products and distillates
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
Specifica
Diametro (sistema metrico) interno 0.89 mm
Materiale PVC (Solvent-ready)
Diametro (sistema metrico) esterno 2.82 mm
Min. Operating Temperature (Metric) °
Quantità confezione 1 Roll
Dimensioni dei tubi 26
Max. Operating Temperature (Metric) 74°C
Product Type Extension Pump Tubing