Learn More
Pyrex™ Graduated Dropping Funnel with Glass Key
Codice prodotto. p-8002579
Click to view available options
Capacità (sistema metrico):
50 mL
100 mL
250 mL
500 mL
1000 mL
Dimensione della confezione:
Pezzo
Codice prodotto. 11333684
Marca: Pyrex 3590/08
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Used to transfer fluids.
  • Complies with ISO 4800 type 4
  • Manufactured from Pyrex borosilicate glass
  • Double graduated scale showing approximate volume contained/dispensed
  • Graduations in white enamel
  • With interchangeable glass key
  • Supplied with glass stopper
TRUSTED_SUSTAINABILITY
Specifica
Capacità (sistema metrico) 1000 mL
Materiale Borosilicate Glass
Graduato Yes
Include Glass stopper and key
Graduazioni 20mL
Product Type Dropping Funnel