Learn More
Pyrex™ Equalizing Glass Graduated Funnel with PTFE Key
Click to view available options
Capacità (sistema metrico):
50 mL
100 mL
250 mL
500 mL
Dimensione della confezione:
Pezzo
Descrizione
- Complies with ISO 4800, Type 4
- With interchangeable PTFE key for grease free performance
- Double graduated scale showing volume of liquid contained/delivered
- Stem inside cone for easy fitting to Quickfit™ assemblies
- Graduations in white enamel
- Supplied with hollow glass stopper
Specifica
Specifica
| Capacità (sistema metrico) | 500 mL |
| Materiale | Borosilicate Glass |
| Graduato | Yes |
| Product Type | Pressure-equalizing Funnel |