Learn More
Pyrex™ Equalizing Glass Graduated Funnel with PTFE Key
Codice prodotto. p-8002582
Click to view available options
Capacità (sistema metrico):
50 mL
100 mL
250 mL
500 mL
Dimensione della confezione:
Pezzo
Codice prodotto. 12905131
Marca: Pyrex 443547
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
For use when liquids have to be added to systems under vacuum.
  • Complies with ISO 4800, Type 4
  • With interchangeable PTFE key for grease free performance
  • Double graduated scale showing volume of liquid contained/delivered
  • Stem inside cone for easy fitting to Quickfit™ assemblies
  • Graduations in white enamel
  • Supplied with hollow glass stopper
TRUSTED_SUSTAINABILITY
Specifica
Capacità (sistema metrico) 250 mL
Materiale Borosilicate Glass
Graduato Yes
Product Type Pressure-equalizing Funnel