Learn More
Corning™ PYREX™ Class A Volumetric Flasks with Glass Standard Taper Stopper
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
:
3 L
Descrizione
The strength of these flasks has been increased appreciably through machine-blown bodies to which are sealed heavy-tubing necks tooled for standard taper glass stoppers. The graduation line is sharp and permanent; large white block letters make the inscription easy to read. The 1 mL and 2 mL sizes are test tube shaped. Sizes 3L through 6L have pennyhead standard taper stoppers.
For stopper only, see Cat. Nos. 7650 and 7661.
Specifica
Specifica
| Materiale chiusura | Vetro |
| Diametro (sistema imperiale) esterno | 7.08 in. |
| Capacità (sistema metrico) | 3000 mL |
| Materiale | Borosilicate Glass |
| Altezza (sistema imperiale) | 17.3 in. |
| Altezza (sistema metrico) | 44 cm |
| Stile collo | Long |
| N. per custodia | 1/Cs. |
| N. per confezione | 1/Pk. |
| Tolleranza | ±1 mL |
| Vedi altri risultati |