Learn More
Pyrex™ Funnel Repair Stopcocks with PTFE Key
Click to view available options
Foro spina:
1.5 mm Key Bore
2.5 mm Key Bore
4 mm Key Bore
6 mm Key Bore
Dimensione della confezione:
pezzi 5
Descrizione
- Contains one flared end for connection to the base of the funnel and 100mm long stem with angled tip
- Straight bore
Specifica
Specifica
| Tipo | Funnel Repair |
| Foro spina | 1.5 mm Key Bore |
| Diametro (sistema metrico) | 12.5 mm |
| Materiale | Glass |
| Include | PTFE key |