Learn More
Grant Instruments™ QR Racks for Grant Heated Circulating Baths
Click to view available options
Capacità (sistema imperiale):
10 tubes
16 Tubes
30 Tubes
35 Tubes
44 Tubes
5 Tubes
Capacità:
0.5 mL Tubes
1.5 mL Tubes
10 to 13 mm Tubes
16 to 19 mm Tubes
24 mm Tubes
30 mm Tubes
Dimensione della confezione:
Pezzo
Descrizione
- Accommodates tubes as small as 0.5mL or as large as 30mm dia.
- For use with Grant Heated Circulating Baths and Heating Circulators.
Specifica
Specifica
| Capacità (sistema imperiale) | 5 Tubes |
| Da utilizzare con (apparecchiatura) | Grant Heated Circulating Baths |
| Capacità | 30 mm Tubes |