Learn More
Applied Biosystems™ QuantStudio™ 12K Flex OpenArray™ Practice Kit
Designed to introduce newcomers to the OpenArray™ block workflow, as well as to provide instrument verification.
Marca: Applied Biosystems™ 4469620
Descrizione
The kit contains 6 practice loading arrays plus reagents. The practice kit also includes these accessories: 6 lids and plugs, 6 immersion fluid syringes, 2 carriers, 1 box of AccuFill™ tips, and 384-well plates.
- PCR Method: qPCR
- Primer-Probe Compatability: TaqMan™
Specifica
| The practice kit includes a QuantStudio™ Accessories Kit, Accufill™ System tips, 384-well sample plates and loading plate, and TaqMan™ OpenArray™ Genotyping Master Mix. | |
| Low | |
| Standard | |
| QuantStudio™ 12k Flex | |
| OpenArray™, QuantStudio™ | |
| DNA |
| Primer-Probe | |
| qPCR | |
| SNP Genotyping (Real-Time PCR-Based) | |
| SNPs (Known) | |
| 12K Flex Practice Kit |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.