Learn More
Hellma™ Quartz Suprasil Match Pair Dish
Click to view available options
Materiale:
Quartz Suprasil
Unit Size:
pezzi 2
Specifications
Specifications
| Tipo | Match Pair Dish |
| Materiale | Quartz Suprasil |
| Quantità | 2Pack |
| Wavelength Range | 200 to 2500 nm |