Learn More
Matraccio di Erlenmeyer in vetro borosilicato Quickfit™ con imboccatura in vetro smerigliato Quickfit
Click to view available options
Capacità (sistema metrico):
5 mL
25 mL
50 mL
100 mL
150 ml
250 mL
500 mL
1000 mL
2000 mL
Altezza (sistema metrico):
54 mm
70 mm
85 mm
90 mm
100 mm
100 mm
116 mm
118 mm
130 mm
130 mm
175 mm
175 mm
220 mm
220 mm
270 mm
Diametro (sistema metrico) esterno:
25 mm
42 mm
51 mm
64 mm
64 mm
74 mm
74 mm
85 mm
85 mm
101 mm
101 mm
130 mm
130 mm
160 mm
Giunto a bussola:
14/23
19/26
24/29
29/32
34/35
Dimensione della confezione:
pezzi 10
pezzi 2
pezzi 20
pezzi 5
Descrizione
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Glass Erlenmeyer Flask with Quickfit Ground Glass Socket complies to ISO 4797.
- Complies to ISO 4797
- Conical flasks with highest quality Quickfit™ ground glass socket
- Borosilicate glass for corrosion free performance and high resistance to thermal shock
- Designed for the estimation of arsenic and antimony
- Method uses the formation of a complex between arsine and silver diethyldithiocarbamate with subsequent measurement at 538nm or 5380.
Specifica
Specifica
| Certificazione/Conformità | ISO 4797 |
| Capacità (sistema metrico) | 500 mL |
| Materiale | Borosilicate Glass |
| Altezza (sistema metrico) | 175 mm |
| Include | Ground socket |
| Forma | Conico |
| Giunto a bussola | 19/26 |
| Diametro (sistema metrico) esterno | 101 mm |