Learn More
Quickfit™ Borosilicate Glass Ether Condenser
Click to view available options
Nessun giunto rastremato standard:
19/26
Dimensione della confezione:
pezzi 2
Descrizione
- Highly efficient coil condenser suitable for condensing very volatile liquids
- Inner coil with large surface area
- Outer water jacket to avoid vapour creep around the coil
- Coil and jacket are independent and can be fed from separate water supplies for maximum efficiency
- With detachable plastic screwthread connectors for the easy and safe attachment of flexible tubing
Specifica
Specifica
| Da utilizzare con (applicazione) | Suitable for condensing very volatile liquids |
| Nessun giunto rastremato standard | 19/26 |
| Materiale | Borosilicate Glass |
| Tipo | Ether Condenser |