Learn More
Quickfit™ Chromatography Columns with Rotaflo™ Stopcock and Ground Cone
Click to view available options
Lunghezza (sistema metrico):
270 mm
Dimensione della confezione:
pezzi 5
Descrizione
- With glass sintered disc (porosity 0) to support column packing
- GP Rotaflo™ stopcock with PTFE key for fine control of eluent flow
- Stem within lower cone allows mounting on a Büchner flask for vacuum flash chromatography
- Manufactured from borosilicate glass
Specifica
Specifica
| Descrizione | Chromatography Column |
| Quantità | 1 Pack |
| Diametro (sistema metrico) interno | 10 mm |
| Lunghezza (sistema metrico) | 270 mm |
| Materiale | Pyrex™ Borosilicate Glass |
| Tipi di connettore | 19/26 Ground cone |
| Include | GP Rotaflo™ stopcock with PTFE key, Glass sintered disc |