Learn More
Quickfit™ Glass Joint Sockets
Codice prodotto. p-8002998
Click to view available options
Joint Size:
10/19
12/21
14/23
19/26
24/29
29/32
34/35
40/38
45/40
60/46
7/16
Spessore (sistema metrico) parete:
1.5mm
1.8mm
2mm
2.3mm
2.5mm
3.2mm
Quantità:
1/Pk.
10/Pk.
5/Pk.
Dimensione della confezione:
pezzi 10
pezzi 5
pezzi 50
Codice prodotto. 10243270
Marca: Quickfit SRB24UB
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Manufactured with minimum shank length of 100mm
  • Unbadged ground glass sockets
  • Free from any ceramic markings
  • Avoids problem of badge charring at high temperatures that can give untidy appearance
  • High quality joints manufactured to accuracy that surpasses requirements of ISO 383
  • Accurate taper and fine surface finish reduce possibility of joint seizure or leakage
  • With round profile bead on top of socket for extra mechanical strength
TRUSTED_SUSTAINABILITY
Specifica
Quantità 10/Pk.
Tipo Glass joint socket
Joint Size 24/29
Spessore (sistema imperiale) parete 0.08 in.
Spessore (sistema metrico) parete 2mm