Learn More
Rondelle filettate in PTFE Quickfit™
Codice prodotto. 10538220
Click to view available options
Dimensioni filo:
13
18
24
28
Diametro (sistema metrico):
10.5 to 11.5 mm (through cap)
6 to 7 mm (through cap)
7 to 8.5 mm (through cap)
8.5 to 9.5 mm (through cap)
Dimensione della confezione:
pezzi 10
Codice prodotto. 10538220
Marca: Quickfit QW18/7
Secure screwcaps to glass screwthreads. QuickFit™ screwthread washers for QuickFit™ screwthreads ensure that the cap fits correctly. Washers come in a variety of sizes.
  • For use with Quickfit™ glass screwthreads
  • Melamine screwcaps with aperture at the top
  • Silicone rubber rings with PTFE washers
  • To determine what components are required: i) Check the size of the glass screw thread (size designation refers to O. D. of thread inmm) ii) Check the O. D. of the sampling tube, thermometer, probe etc. , that is to be passed through the screw cap.
TRUSTED_SUSTAINABILITY
Specifica
Diametro (sistema metrico) 7 to 8.5 mm (through cap)
Materiale PTFE
Da utilizzare con Per filettature in vetro Quickfit™
Dimensioni filo 18
Product Type Rondella