Learn More
Adattatore presa/cono Quickfit™
Click to view available options
Cone Size:
14/23
19/26
24/29
29/32
34/35
NS 24/29
N. giunto rastremato standard:
14/23
19/26
24/29
29/32
NS 24/29
Dimensione della confezione:
Pezzo
pezzi 5
Descrizione
Quickfit™ Tubing Adapter with Side-arm for attachment of rubber hose
- With sidearm for the attachment of rubber hose etc
- Detachable plastic screwthread connector allows flexible tubing to be fitted easily and safely
Specifica
Specifica
| Cone Size | 19/26 |
| Include | Detachable Plastic screw thread connector |
| Materiale | Borosilicate Glass |
| Product Type | Side-arm tubing adapter |
| N. giunto rastremato standard | 14/23 |