Learn More
Quickfit™ Spherical Ball Joints, Male
Click to view available options
Joint Size:
S13
S19
S29
S35
S41
Spessore (sistema metrico) parete:
1.5mm
2.5mm
2.8mm
Dimensione della confezione:
Pezzo
pezzi 10
Descrizione
Intended for use with female spherical cup joints FS:
- Made from Pyrex borosilicate glass
- Manufactured with an approximate shank length of 100mm
Specifica
Specifica
| Tipo | Male Spherical Joint |
| Joint Size | S29 |
| Spessore (sistema metrico) parete | 2.5mm |