Learn More
Quickfit™ Unbadged Joints Cones
Codice prodotto. 12939363
Click to view available options
Cone Size:
10/19
12/21
14/23
19/26
24/29
29/32
34/35
40/38
45/40
55/44
7/16
Spessore (sistema metrico) parete:
1mm
1.5mm
1.8mm
2.00mm
2mm
2.30mm
5mm
Dimensione della confezione:
pezzi 10
pezzi 20
pezzi 5
pezzi 50
Codice prodotto. 12939363
Marca: Quickfit CNB10UB
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Free from any ceramic marking
  • Avoids problem of badge charring at high temperatures which can give an untidy appearance
  • Manufactured to accuracy which surpasses requirements of ISO 383
  • Pyrex™ borosilicate glass for resistance to chemical corrosion and thermal shock
  • Cones manufactured with shank length of 100mm
TRUSTED_SUSTAINABILITY
Specifica
Tipo Joint Cone
Cone Size 10/19
Spessore (sistema metrico) parete 1mm