Learn More
(R)-2-(Boc-amino)butyric acid, 95%
Codice prodotto. p-7055415
Click to view available options
Quantità:
1 g
5 g
25 g
Dimensione della confezione:
1g
25g
5g
Codice prodotto. 15440999
Marca: Thermo Scientific Alfa Aesar H63991.03
Richiesta di formato massivo o personalizzato
CAS: 45121-22-0 | C9H17NO4 | 203.238 g/mol
(R)-2-(Boc-amino)butyric acid is used in coupling reaction. Also used in preparation of dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(R)-2-(Boc-amino)butyric acid is used in coupling reaction. Also used in preparation of dyes.
Solubility
Soluble in DMF (1mmol in 1ml DMF).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 45121-22-0
Formula molecolare C9H17NO4
Molecular Weight (g/mol) 203.238
Numero MDL MFCD00270335
InChI Key PNFVIPIQXAIUAY-ZCFIWIBFSA-N
Sinonimo boc-d-abu-oh, r-2-tert-butoxycarbonyl amino butanoic acid, r-n-boc-2-aminobutyric acid, boc-d-alpha-aminobutyric acid, boc-r-2-aminobutyric acid, boc-d-2-aminobutyric acid, 2r-2-tert-butoxycarbonyl amino butanoic acid, 2r-2-tert-butoxy carbonyl amino butanoic acid, butanoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2r, 2-r-tert-butoxycarbonylaminobutyric acid
PubChem CID 2755936
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CCC(C(=O)O)NC(=O)OC(C)(C)C
Specifica
Materiale o nome chimico (R)-2-(Boc-amino)butyric acid
Colore Yellow
Quantità 1 g
Informazioni di solubilità Soluble in DMF (1mmol in 1ml DMF).
Peso formulazione 203.24
Percent Purity 95%
Forma fisica Liquid