Learn More
R&D Systems™ Cultrex Basement Membrane Extract, Pathclear (w/ Phenol Red)
Click to view available options
Quantità:
5 mL
2 x 5 mL
Dimensione della confezione:
10mL
5mL
Descrizione
Functional Assay: Tube formation - BME promotes formation of capillary-like structures by human (HBMVEC; HUVEC) or mouse (SVEC4-10) endothelial cells. Gelling Assay: BME gels in less than 30 minutes at 37degreesC, and maintains the gelled form in culture medium for a minimum of 14 days at 37degreesC.
Specifica
Specifica
| Concentrazione | 12 to 18 mg/mL |
| Da utilizzare con (applicazione) | Bioactivity |
| Quantità | 5 mL |
| Livello di endotossine | ≤8EU/mL |
| Sorgente | Murine Engelbreth-Holm-Swarm (EHS) tumor. |
| Content And Storage | Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles. |