Learn More
R&D Systems™ Recombinant Human MMP-3 Western Blot Standard Protein
Codice prodotto. 18147640
Click to view available options
Quantità:
1 Vial
Dimensione della confezione:
100µL
Codice prodotto. 18147640
Marca: R&D Systems™ WBC015
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Western Blot
The Recombinant Human MMP-3 Western Blot Standard Protein has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Specifica
Da utilizzare con (applicazione) Western Blot
ID gene (immissione) 4314
Nome MMP-3 Western Blot Standard
Quantità 1 Vial
Requisiti di stoccaggio Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Simbolo del gene MMP3
Coniugato Unconjugated
Recombinant Recombinant
Proteine MMP-3