Learn More
R&D Systems™ Recombinant Human MMP-7 Western Blot Standard Protein
Click to view available options
Quantità:
1 Vial
Dimensione della confezione:
100µL
Descrizione
The Recombinant Human MMP-7 Western Blot Standard Protein has been validated for the following applications: Western Blot.
Specifica
Specifica
| Da utilizzare con (applicazione) | Western Blot |
| ID gene (immissione) | 4316 |
| Nome | MMP-7 Western Blot Standard |
| Quantità | 1 Vial |
| Requisiti di stoccaggio | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Simbolo del gene | MMP7 |
| Coniugato | Unconjugated |
| Recombinant | Recombinant |
| Proteine | MMP-7 |