Learn More
R&D Systems™ Sialyltransferase Activity Kit
Click to view available options
Quantità:
1 Kit
Dimensione della confezione:
1 set
Descrizione
Includes
CHO cell-expressed Recombinant Human CD73, CMP, Phosphatase Buffer 2, 100mM MnCl2, Phosphate Standard, Malachite Green Reagents A and B
Uses the 5 ft.-nucleotidase CD73 as a coupling phosphatase to remove inorganic phosphate quantitatively from the leaving nucleotide cytidine 5 ft.-monophosphate (CMP) that is generated during sialyltransferase reactions
No t
Specifica
Specifica
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantità | 1 Kit |