Learn More
R&D Systems™ Mouse Bradykinin RB2/BDKRB2 (NP_033877) VersaClone cDNA
Codice prodotto. 18788458
Click to view available options
Quantità:
10 μg
Dimensione della confezione:
10µg
Codice prodotto. 18788458
Marca: R&D Systems™ RDC1120
Bradykinin Receptor B2
The Mouse Bradykinin RB2/BDKRB2 (NP_033877) VersaClone cDNA clone contains the complete ORF for Bradykinin RB2/BDKRB2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Bradykinin RB2/BDKRB2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
TRUSTED_SUSTAINABILITY
Specifica
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantità 10 μg
For Reasearch Use Only; Not for Use in Humans