Learn More
R&D Systems™ Mouse Bradykinin RB2/BDKRB2 (NP_033877) VersaClone cDNA
Click to view available options
Quantità:
10 μg
Dimensione della confezione:
10µg
Descrizione
The Mouse Bradykinin RB2/BDKRB2 (NP_033877) VersaClone cDNA clone contains the complete ORF for Bradykinin RB2/BDKRB2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Bradykinin RB2/BDKRB2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
Specifica
Specifica
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantità | 10 μg |
For Reasearch Use Only; Not for Use in Humans