Learn More
ALDH8A1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ALDH8A1.
Marca: Abnova PAB3143.400uL
Descrizione
This protein belongs to the aldehyde dehydrogenases family of proteins. It plays a role in a pathway of 9-cis-retinoic acid biosynthesis in vivo. This enzyme converts 9-cis-retinal into the retinoid X receptor ligand 9-cis-retinoic acid, and has approximately 40-fold higher activity with 9-cis-retinal than with all-trans-retinal. Therefore, it is the first known aldehyde dehydrogenase to show a preference for 9-cis-retinal relative to all-trans-retinal. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq
Specifica
| ALDH8A1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ALDH8A1. | |
| In PBS (0.09% sodium azide) | |
| NP_072090;Q9H2A2 | |
| ALDH8A1 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human ALDH8A1. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| ALDH8A1 | |
| ALDH12/DJ352A20.2/DKFZp779D2315/MGC138650 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 64577 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only