missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Canine IgM Heavy Chain Secondary Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-73845-10MG
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgM Heavy Chain Polyclonal specifically detects IgM Heavy Chain in Canine samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Specifica
| IgM | |
| Polyclonal | |
| FITC | |
| Lyophilized from 0.02M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2, with 10Mg/ml Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide | |
| Rabbit | |
| Ion exchange chromatography | |
| Secondary | |
| Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum, Dog IgM and Dog Serum. No reaction was observed against Dog IgG. | |
| Canine | |
| Purified |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| LYOPH | |
| Flow Cytometry 1:500 - 1:2500, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
| AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
| Dog IgM mu heavy chain | |
| 10 mg | |
| 3507 | |
| Reconstitute with 2.0mL of deionized water. | |
| Store at 4°C in the dark. | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto