Learn More
Rabbit anti-Chicken IgG whole molecule, HRP, Polyclonal, Secondary Antibody, Abnova™
Marca: Abnova PAB10506.1500ug
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
| Rabbit anti-Chicken IgG whole molecule | |
| Polyclonal | |
| Rabbit anti-chicken IgG recognizes chicken IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Peroxidase. | |
| Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% gentamicin sulfate) | |
| Chicken IgG whole molecule | |
| 1500 μg | |
| Chicken | |
| Lyophilized |
| Dot Blot, ELISA, Immunohistochemistry, Western Blot | |
| HRP | |
| The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
| Secondary | |
| Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Titolo del prodotto
For Research Use Only