missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Goat IgG Fc Secondary Antibody, HRP (Pre-adsorbed), Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-74864
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgG Fc Polyclonal antibody specifically detects IgG Fc in Goat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifica
| IgG Fc | |
| Polyclonal | |
| HRP | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Based on IEP, this antibody reacts with heavy gamma chains on goat IgG. This antibody has been pre-adsorbed against human serum proteins. | |
| Goat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot 1:200-1:5000, ELISA 1:200-1:5000, Immunohistochemistry 1:500-1:5000, Immunohistochemistry-Paraffin | |
| Purified Goat IgG Fc fragment | |
| 1 mg | |
| Secondary | |
| Rehydrate with 1.1 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. | |
| Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto