missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Guinea pig IgG F(c) fragment, Biotin, Polyclonal, Secondary Antibody, Abnova™
Marca: Abnova PAB10641.2mg
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
| Rabbit anti-Guinea pig IgG F(c) fragment | |
| Polyclonal | |
| Rabbit anti-guinea pig IgG recognizes Guinea pig IgG F(c) fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Biotin. | |
| Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| Guinea pig IgG F(c) fragment | |
| 2 mg | |
| Guinea Pig | |
| Lyophilized |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| Biotin | |
| The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
| Secondary | |
| Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto