missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Monkey IgA Fc Secondary Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-69197
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
IgA Fc Polyclonal antibody specifically detects IgA Fc in Monkey samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifikationer
| IgA Fc | |
| Polyclonal | |
| FITC | |
| Lyophilized from PBS (pH 7.2) | |
| Pools of purified IgA isolated from Rhesus monkey serum. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
| 1 mL | |
| Secondary | |
| Monkey | |
| Lyophilized |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| LYOPH | |
| ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen | |
| Rabbit | |
| Ion exchange chromatography | |
| RUO | |
| Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C. | |
| Store at 4°C in the dark. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering