missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgG F(c) fragment, FITC, Polyclonal, Secondary Antibody, Abnova™
Marca: Abnova PAB10775.1500ug
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
| Rabbit anti-Mouse IgG F(c) fragment | |
| Polyclonal | |
| Rabbit anti-mouse IgG recognizes mouse IgG F(c) fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Fluorescein. | |
| Lyophilized from 10mM sodium phosphate, 140mM NaCl, pH 7.4 (0.01% (w/v) thimerosal, 10mg/mL BSA (Immunoglobulin and Protease free)) | |
| Mouse IgG F(c) fragment | |
| 1500 μg | |
| Mouse | |
| Lyophilized |
| Flow Cytometry, Immunofluorescence | |
| FITC | |
| The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
| Secondary | |
| Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto